missing translation for 'onlineSavingsMsg'
Learn More

Tenak™ Eco AluRack for Chest Freezer, for Microtiterplates, 65 mm Shelf Height

Eco AluRack for chest freezers is designed to hold microtiterplates.

Brand:  Tenak™ TE15192

Additional Details : Weight : 1.60000kg

 View more versions of this product

Product Code. 12468190

  • 67.00 EUR / Each
Estimated Shipment: 02-08-2024
to see stock.
Add to basket

Description

Description

  • For use in chest freezers
  • Racks can either be used as one plate per shelf or the plates can be stacked
  • Constructed of aluminum
Specifications

Specifications

Microtiterplates (5)
Chest Frseezers
65 mm
Aluminum
135 x 92 x 343 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers