Learn More
Tenak™ Eco AluRack for Chest Freezer, for Microtiterplates, 16 mm Shelf Height
Eco AluRack for chest freezers is designed to hold microtiterplates.
Marca: Tenak™ TE15102
Dettagli aggiuntivi : Weight : 1.60000kg
Descrizione
- For use in chest freezers
- Racks can either be used as one plate per shelf or the plates can be stacked
- Constructed of aluminum
Specifica
Microtiterplates (19) | |
Chest Frseezers | |
16 mm |
Aluminum | |
135 x 92 x 334 mm |