Learn More
BTX™ ECM™ 2001+ HV Output Cable
ECM™ 2001+ HV Output Cable for use with BTX™ ECM™ 2001+.
Brand: BTX™ 452058
Additional Details : Weight : 0.30000kg
Description
- For connection to electroporation generators
Specifications
In-Vivo Drug or Gene Delivery, Gene Therapy, Protein Delivery | |
Cables and Adapters | |
Cable Only | |
3 m | |
Non-sterile | |
AC Sine Wave, DC Square Wave |
ECM™ 2001+ | |
ECM 2001+ HV Output Cable, 10 ft | |
10 ft. | |
1 Ea. | |
Output Cable |