Learn More
Ebro Electronic™ EBI 100-TP230 Temperature/Pressure Loggers
For monitoring and validation of processes in steam stabilizers, hot air stabilizers, incubators, refrigerators, climate cabinets and more. Ebro Electronic™ EBI 100-TP230 Temperature/Pressure Loggers are provided with a rigid metal probe.
Brand: Ebro Electronic™ 13406554
Additional Details : Weight : 0.80000kg
Description
- F-value and A0 value calculation
- Without radio technology
- Temperature resistant up to 150°C
- High accuracy
- Completely waterproof
- Programming and evaluation with PC
- Exchangeable battery
- Acc. to EN 285, ISO 17665 and ISO 15883
Specifications
EBI 100-TP230 Temperature/Pressure Logger | |
Lithium | |
IP 68 | |
Pt 1000 | |
±0.3°C | |
0°C to 150°C |
2 x 9000 Readings | |
Calibration certificate | |
EBI 100-TP230 | |
Monitoring and validation of processes in steam sterilizers, hot air sterilizers, incubators, refrigerators, climate cabinets | |
0.1°C | |
70 g |