Learn More
Ebro Electronic™ EBI 10-T230 Temperature Data Logger
Brand: Ebro Electronic™ 13406106
Additional Details : Weight : 0.80000kg
Specifications
Datalogger, EBI 10-T230 Temperature | |
2 Years | |
IP 68 / NEMA 6 | |
Pt 1000 | |
0.01°C | |
50 mm |
100,000 Readings | |
Factory Calibration certificate | |
EBI 10-T230 | |
±0.5 (–85 to-40) ±0.2 (–40 to 0) ±0.1 (0 to 140 ) ±0.2 (140 to150)°C | |
-85°C to +150°C | |
70 g |