Learn More
Ebro Electronic™ EBI 10-T100 Temperature Data Loggers with Internal Sensors
Provides high accuracy in process monitoring. Ebro Electronic™ EBI 10-T1000 Temperature Data Loggers with Internal Sensors are temperature resistant up to 150°C.
Brand: Ebro Electronic™ 1340-6100
Additional Details : Weight : 0.07000kg
Description
- Completely waterproof IP 68
- Programming and evaluation with PC
- Exchangeable battery
Specifications
Datalogger, EBI 10-T100 Temperature | |
3.6 V | |
1 | |
IP 68 / NEMA 6 | |
Pt 1000 | |
±0.5 (–85 to -40) ±0.2(–40 to 0) ±0.1 (0 to 140) ±0.2(140 to 150)°C | |
-85°C to +150°C |
100,000 Readings | |
2 Years | |
Factory calibration certificate | |
EBI 10-T100 | |
For measurements in the beverage industry, Bottle cleaning, Core temperature measurement | |
0.01°C | |
70 g |