Learn More
Cole-Parmer™ Dye Tablets
Cole-Parmer Dye Tablets, Yellow/Green, Dye Tracer, For Use With: Plumbing, storm and sanitary drain analysis, Quantity: 200/Pk.
66.55 EUR - 67.95 EUR
Specifications
For Use With (Application) | Plumbing, storm and sanitary drain analysis |
---|---|
Quantity | 200/Pk. |
Type | Dye Tracer |
Product Code | Brand | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Price | Quantity & Availability | |||||
11747947
|
Cole-Parmer™
0029516 |
Dark Red |
67.95 EUR
Pack of 200 |
Estimated Shipment: 12-07-2024
Log in to see stock. |
|||||
11757947
|
Cole-Parmer™
0029517 |
Yellow/Green |
66.55 EUR
Pack of 200 |
Estimated Shipment: 12-07-2024
Log in to see stock. |
|||||
Description
Cole-Parmer Dye Tablets, Yellow/Green, Dye Tracer, For Use With: Plumbing, storm and sanitary drain analysis, Quantity: 200/Pk.
Specifications
Plumbing, storm and sanitary drain analysis | |
200/Pk. | |
Dye Tracer |