Learn More
Macherey-Nagel™ Durasil-25 TLC Plate
181.00 EUR - 203.00 EUR
Specifications
Material | Durasil-25 |
---|---|
Particle Size | 5 to 17 μm |
Pore Size | 60 Å |
Type | TLC Plate |
Pore Volume | 0.75 mL/g |
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | |||||
10318171
|
Macherey-Nagel™
812003 |
10 x 20 cm |
203.00 EUR
Pack of 50 |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
10490702
|
Macherey-Nagel™
812004 |
20 x 20 cm |
181.00 EUR
Pack of 25 |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
Specifications
Durasil-25 | |
60 Å | |
0.75 mL/g | |
0.25 mm |
5 to 17 μm | |
TLC Plate | |
∼500 m2/g |