Learn More
Lenz Laborglasinstrumente™ Gas Washing Bottle Heads, with Filter Disc
Provides an inexpensive but effective method for the washing and drying of gases. Lenz Laborglasinstrumente™ Gas Washing Bottle Heads, with Filter Disc are manufactured from durable DURAN™ borosilicate glass.
Brand: Lenz Laborglasinstrumente™ 05510400
Additional Details : Weight : 0.10000kg
Description
- Acc. to DIN 12596, with cone
- For bottles acc. to DIN 12463
- Available in a range of porosities
- Corrosion-free and resistant to thermal shock
Specifications
Duran Tubing | |
Filter disc | |
NS 29/32 | |
1 Pack |
Gas Washing Bottle Head with Filter Disc | |
0 | |
DIN 12596, DIN 12463 |