Learn More
DWK Life Sciences DURAN™ Reduction Adapter, socket/cone
Reduction from sleeve to core.
Brand:  DWK Life Sciences 286535906
149.15 EUR valid until 2024-12-31
Use promo code "23258" to get your promotional price.
Additional Details : Weight : 0.20000kg
 View more versions of this product
Product Code. 15961601
  • 157.00 EUR / Pack of 10
Estimated Shipment: 22-05-2024
Description
Adapters are used to connect two components with different NS sizes. DURAN™ reducers allow the connection of a larger sleeve to a smaller core.
  • Reducers allow the connection of a larger sleeve to a smaller core and thus the use of differently sized ground joint components.
Specifications
Specifications
10/Pk.
Connection of two components with different standard ground joint sizes
Socket NS 19/26, Cone NS 14/23