Learn More
DWK Life Sciences DURAN™ Hexagonal Stopper, pointed, diamond-ground
Hollow glass stopper for sealing with draining tip.
1.09 EUR - 312.00 EUR
Specifications
Autoclavable | Yes |
---|---|
Material | Boro 3.3 |
For Use With (Application) | Closure of vessels with standard ground joints |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
11772185
|
DWK Life Sciences
286116608 |
NS 24/29 |
251.00 EUR
Pack of 100 |
Estimated Shipment: 17-04-2024
Log in to see stock. |
|||||
15864548
|
DWK Life Sciences
286115406 |
NS 14/23 |
169.00 EUR
Pack of 100 |
Estimated Shipment: 30-04-2024
Log in to see stock. |
|||||
15911521
|
DWK Life Sciences
286117407 |
NS 29/32 |
312.00 EUR
Pack of 100 |
Estimated Shipment: 08-05-2024
Log in to see stock. |
|||||
15921521
|
DWK Life Sciences
286117904 |
NS 34/35 |
89.40 EUR
Pack of 10 |
Estimated Shipment: 02-05-2024
Log in to see stock. |
|||||
15884548
|
DWK Life Sciences
286118103 |
NS 45/40 |
128.00 EUR
Pack of 10 |
Estimated Shipment: 02-05-2024
Log in to see stock. |
|||||
15874548
|
DWK Life Sciences
286116008 |
NS 19/26 |
222.00 EUR
Pack of 100 |
Estimated Shipment: 17-04-2024
Log in to see stock. |
|||||
15854548
|
DWK Life Sciences
286113808 |
NS 7/16 |
263.00 EUR
Pack of 100 |
Estimated Shipment: 17-04-2024
Log in to see stock. |
|||||
10621872
|
DWK Life Sciences
286114804 |
NS 12/21 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10365191
|
DWK Life Sciences
286114204 |
NS 10/19 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
DURAN™ hexagonal hollow plugs are used for sealing laboratory equipment such as volumetric flasks, mixing cylinders, ground joint flasks, etc. DURAN™ hexagonal hollow plugs are available in 2 standard versions - pointed - flat - Sizes analogous to DURAN™ conical ground joints. Cut quality: DG = Diamond Ground
- Pointed
- Diamond-ground
- With a dripping syringe, for example, a condensate can be collected and the reaction medium recirculated.
- hollow glass stopper
Specifications
Yes | |
Boro 3.3 | |
Closure of vessels with standard ground joints |