Learn More
Lenz Laborglasinstrumente™ Duran Glass Absaugstück
Brand: Lenz Laborglasinstrumente™ B0292561
Additional Details : Weight : 0.02700kg
Specifications
Suction Piece | |
Duran Glass | |
1Each | |
NSK 29/32 |