Learn More
DWK Life Sciences DURAN™ Gas-washing Bottle, lower part, Drechsel type with NS 29/32, height 200 mm
For cleaning gases.
Brand: DWK Life Sciences 247144405
Additional Details : Weight : 0.40800kg
Description
DURAN™ gas washing bottle bases can be completed with various attachments to form complete washing bottles.
- After turning with NS 29/32, height 200 mm
- According to DIN 12463
- Can be completed to complete wash bottles with various attachments
Specifications
500 mL | |
Borosilicate Glass (3.3) | |
Purification of gases | |
10/Pk. |