Learn More
DWK Life Sciences DURAN™ Funnel, ribbed
A funnel enables a clean transfer and filtration of liquids.
Brand: DWK Life Sciences 213524104
296.40 EUR valid until 2024-12-31
Use promo code "23258" to get your promotional price.
Additional Details : Weight : 0.70000kg
Description
Transferring liquids and solids is a common working method. In this activity the funnel is used, which due to its shape can absorb a relatively large amount of liquid or solid and transfer it through a constriction into a vessel with a small opening. Funnels are available in different sizes and designs, selected according to their application.
- A sticking to a smooth glass surface and thus a slowing down of the filtration speed is prevented by the ribbed shape
- The ribbed form is ideal for filtering with round-paper
- Fired-on, highly durable white print
Specifications
80 mm | |
Yes | |
10/Pk. |
Boro 3.3 | |
Filtering of liquids; The spillage of substances is avoided; Filtering |