Learn More
DWK Life Sciences DURAN™ Filter Tube, Allihn type
Micro filter tubes can be used to filter very small quantities of solution. The Allihn design has a large volume (30 ml) compared to other microfilters.
400.00 EUR - 434.00 EUR
Specifications
Material | Borosilicate Glass (3.3) |
---|---|
For Use With (Application) | Filter design according to Allihn; Filtration of very small quantities; Filtration at reduced pressure; Qualitative inorganic analysis and preparative chemistry; Filtering |
No. per Pack | 10/Pk. |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
12526548
|
DWK Life Sciences
258532206 |
Porosity 2 |
400.00 EUR
Pack of 10 |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
15262049
|
DWK Life Sciences
258532309 |
Porosity 3 |
400.00 EUR
Pack of 10 |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
15272049
|
DWK Life Sciences
258532403 |
Porosity 4 |
434.00 EUR
Pack of 10 |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
15252049
|
DWK Life Sciences
258532103 |
Porosity 1 |
400.00 EUR
Pack of 10 |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
Description
One of the most important separation methods in the laboratory environment is filtration, which is used to separate or clean different media. For this working method, filters are required which are selected according to the application with regard to size and porosity. Filter funnels are filters that are used for solid-liquid separation by means of vacuum. Filter frits have a glass sinter bottom which serves as a filter medium and does not require any further filter paper.
- Allihn type
- Suitable for filtration of very small quantities due to filter size
- Fired-on, highly durable white print
Specifications
Borosilicate Glass (3.3) | |
Filter design according to Allihn; Filtration of very small quantities; Filtration at reduced pressure; Qualitative inorganic analysis and preparative chemistry; Filtering | |
10/Pk. |