Learn More
DWK Life Sciences DURAN™ Capillary-Single way Stopcock, complete with PTFE key
Shut-off with PTFE plug for grease free operation.
1420.00 EUR - 1595.00 EUR
Specifications
Material | Borosilicate Glass (3.3), PTFE |
---|---|
Autoclavable | Yes |
No. per Pack | 50/Pk. |
For Use With (Application) | Installation in glass apparatus and devices |
Product Code | Brand | Closure Size | Plug Bore | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Closure Size | Plug Bore | Price | Quantity & Availability | ||||
15804718
|
DWK Life Sciences
286411604 |
12.5 NS | 2 mm |
1420.00 EUR
Pack of 50 |
Estimated Shipment: 04-06-2024
Log in to see stock. |
||||
15824718
|
DWK Life Sciences
286412403 |
14.5 NS | 2.5 mm |
1595.00 EUR
Pack of 50 |
Estimated Shipment: 04-06-2024
Log in to see stock. |
||||
Description
DURAN™ single way stopcocks complete with PTFE valve plugs allow grease free operation. The 3-part tap safety catch secures the tap plug in the tap sleeve.
- Polished shut-off valve sleeve with PTFE valve plug for grease-free operation
- The 3-part tap safety catch secures the tap plug in the tap sleeve.
- Taper 1:10
- Glass articles for further processing
Specifications
Borosilicate Glass (3.3), PTFE | |
Yes | |
50/Pk. | |
Installation in glass apparatus and devices |