Learn More
DWK Life Sciences DURAN™ Burette Stopcock, straight, complete with PTFE key
For attaching to burettes.
Brand: DWK Life Sciences 286408208
1249.25 EUR valid until 2024-12-31
Use promo code "23258" to get your promotional price.
Additional Details : Weight : 12.50000kg
Description
Straight DURAN™ burette taps complete with PTFE plug for attachment to burettes have no calibrated tips.
- Straight burette tap sleeve with PTFE plug
- Tip not calibrated
- Grease-free working
- Glass articles for further processing
Specifications
2 mm | |
Borosilicate Glass (3.3), PTFE | |
12.5 NS | |
50/Pk. |
8 mm | |
Yes | |
1.5 mm | |
For attaching to burettes |