Learn More
DWK Life Sciences DURAN™ Burette Stopcock, angled, complete with PTFE key
For attaching to burettes.
Brand: DWK Life Sciences 286409607
1273.00 EUR valid until 2024-12-31
Use promo code "23258" to get your promotional price.
Additional Details : Weight : 5.00000kg
Description
Lateral DURAN™ burette taps complete with PTFE tap plugs for attachment to burettes have no calibrated tips.
- Side burette tap case with PTFE plug
- Tip not calibrated
- Grease-free working
- Glass articles for further processing
Specifications
2.5 mm | |
Borosilicate Glass (3.3), PTFE | |
12.5 NS | |
50/Pk. |
8 mm | |
Yes | |
1.5 mm | |
For attaching to burettes |