Learn More
Lenz Laborglasinstrumente™ DURAN™ Borosilicate Glass Round Bottom Flasks with Conical Ground Joint
Brand:  Lenz Laborglasinstrumente™ 03002976
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10461083
  • 145.22 EUR / Each
Estimated Shipment: 14-06-2024
Specifications
According to DIN EN ISO 4797 and in addition to standard
Borosilicate Glass, Duran Glass
Round
3000 mL
29/32 NS Socket
185 mm