Learn More
Dräger™ Short-Term Detector Tubes - Nitric Acid
Brand: Dräger™ 6728311
Additional Details : Weight : 0.07740kg
Description
1Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.
2Safety Equipment Institute (SEI) certified.
Specifications
Nitric Acid 1/a | |
1 to 50ppm | |
10 | |
41°F to 104°F |
3 to 15mg/L | |
10/20 | |
2 to 4 min. | |
5° to 40°C |