Learn More
Miele Professional™ Dosing system
Dispenser module DOS K 85 for 5-10 litre canisters with level indicator for the automatic dispensing of liquid cleaning agents (PG 8583/93)
Brand: Miele Professional™ 9961070
Additional Details : Weight : 3.00000kg
Description
- Dispenser monitoring – in accordance with DIN EN ISO 15883
- For dispensing liquid cleaning agent or neutralising agent
- For dispensing chemical disinfectant
- 333 mm siphon for 5-10 l containers
- Conversion kit for siphons for 10-30 l canisters available
Specifications
Dispenser | |
5-10 litre canisters |