Learn More
Donkey anti-Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody, FITC, Invitrogen™
Donkey Polyclonal Secondary Antibody
Brand: Invitrogen 31568
122.67 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Additional Details : Weight : 0.80000kg
Description
Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Product # 31568 has been successfully used in Western blot, IF, ICC, IHC, IP and FACS applications. Product # 31568 reacts with the heavy chains of rabbit IgG and with the light chains common to most rabbit immunoglobulins, but does not react against non-immunoglobulin serum proteins. This antibody has been tested by ELISA and/or solid-phase adsorbed to ensure minimal cross-reaction with bovine, chicken, goat, guinea pig, hamster, horse, human, mouse,rat and sheep serum proteins. However, this antibody may cross-react with immunoglobulins from other species. Store product protected from light at 4°C until opened. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C. Fluorescein Amax= 492 nm; Emax= 520 nm. Fluorophore/Protein: 8.3 μg/mg; >3 moles FITC per mole IgG. (lot-dependent). Reconstitute with 0.4 mL of distilled water (1.5 mg/mL after restoration). Country of Origin: USA
Rabbit IgG (H+L) Cross-Adsorbed Polyclonal antibody specifically detects Rabbit IgG (H+L) Cross-Adsorbed in Rabbit samples. It is validated for Immunocytochemistry, Immunohistochemistry, Immunoprecipitation.Specifications
Rabbit IgG (H+L) Cross-Adsorbed | |
Polyclonal | |
FITC | |
Donkey | |
500 μg | |
Secondary | |
4° C, store in dark | |
IgG |
Immunocytochemistry, Immunohistochemistry, Immunoprecipitation | |
1.5 mg/mL | |
PBS with 15mg/mL BSA and 0.05% sodium azide; pH 7.6 | |
Antigen affinity chromatography | |
RUO | |
Rabbit | |
Lyophilized |