Learn More
DNAJC17 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA559349
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: AVVEFATVKA AELAVQNEVG LVDNPLKISW LEGQPQDAVG RSHSGLSKGS VLSERDYESL VMMRMRQAAE RQQLIARMQQ EDQ Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 84%.
DNAJC17 Polyclonal antibody specifically detects DNAJC17 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
DNAJC17 | |
Polyclonal | |
Unconjugated | |
DNAJC17 | |
1700025B16Rik; C87112; D9Bwg1371e; DnaJ (Hsp40) homolog, subfamily C, member 17; DnaJ heat shock protein family (Hsp40) member C17; dnaJ homolog subfamily C member 17; Dnajc17; RGD1308015 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
311329, 55192, 69408 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
D3ZSC8, Q91WT4, Q9NVM6 | |
DNAJC17 | |
Recombinant protein corresponding to Human DNAJC17 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Research Use Only