missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DMPK Recombinant Human Protein
DMPK is a serine/threonine kinase expressed in various muscle and nerve tissues.
Brand: Invitrogen™ PR7186A
Additional Details : Weight : 0.01000kg
Description
Mutations in the untranslated region of the DMPK gene are a characteristic of myotonic dystrophy.
Specifications
NP_004400 | |
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
100μg | |
RUO - research use only | |
DMPK, DM | |
Human | |
GST Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
96.8kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
DMPK Kinase Family | |
Recombinant | |
Baculovirus |