missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DMPK Recombinant Human Protein

DMPK is a serine/threonine kinase expressed in various muscle and nerve tissues.

1826.00 EUR - 7890.00 EUR

Specifications

Accession Number NP_004400
For Use With (Application) Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology
Formulation 50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
Molecular Weight (g/mol) 96.8kDa
Storage Requirements Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Mehr Spezifikationen ansehen

Produkte 2
Produktcode Marke Quantity Preis Menge & Verfügbarkeit  
Produktcode Marke Quantity Preis Menge & Verfügbarkeit  
15458533
missing translation for 'viewDocuments' Aktionsdetails
Invitrogen™
PR7186A
100μg
1826.00 EUR
100µg
Alternative Produkte anzeigen
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 03-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
10392253
missing translation for 'viewDocuments' Aktionsdetails
Invitrogen™
PV3996
1mg
7890.00 EUR
1mg
Alternative Produkte anzeigen
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 03-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Beschreibung

Beschreibung

Mutations in the untranslated region of the DMPK gene are a characteristic of myotonic dystrophy.

Spezifikation

Spezifikation

NP_004400
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
DMPK Kinase Family
Recombinant
Baculovirus
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology
96.8kDa
RUO - research use only
DMPK, DM
Human
GST Tag
Frozen
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate