missing translation for 'onlineSavingsMsg'
Learn More

DL-Homoserine, 98%, Thermo Scientific Chemicals

Catalog Number 15459628
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
1927-25-9
C4H9NO3
119.12
MFCD00002618,MFCD00077786
UKAUYVFTDYCKQA-VKHMYHEASA-N
dl-homoserine, homoserine, h-dl-hoser-oh, d,l-homoserine, h-d-homoser-oh, butyric acid, 2-amino-4-hydroxy-, dl, 2-amino-4-hydroxy-butanoic acid, dl-2-amino-4-hydroxybutyric acid, d-homoserin, h-dl-hse-oh
779
CHEBI:30653
2-amino-4-hydroxybutanoic acid
N[C@@H](CCO)C(O)=O
This item is not returnable. View return policy
1927-25-9
C4H9NO3
119.12
MFCD00002618,MFCD00077786
UKAUYVFTDYCKQA-VKHMYHEASA-N
dl-homoserine, homoserine, h-dl-hoser-oh, d,l-homoserine, h-d-homoser-oh, butyric acid, 2-amino-4-hydroxy-, dl, 2-amino-4-hydroxy-butanoic acid, dl-2-amino-4-hydroxybutyric acid, d-homoserin, h-dl-hse-oh
779
CHEBI:30653
2-amino-4-hydroxybutanoic acid
N[C@@H](CCO)C(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 188°C to 189°C
Beilstein 1721682
Merck Index 14,4739
Formula Weight 119.12
Percent Purity 98%
Quantity 250 mg
Physical Form Solid
Chemical Name or Material DL-Homoserine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'