Learn More
3M™ Disposable Maintenance-Free Half Mask Respirator, 4000 Series
Offers reliable respiratory protection against a range of gases, vapors, and particulates. The 3M Disposable Maintenance-Free Half Mask Respirator can be used until damaged, particulate filters clog, gas filters become saturated or after one month.
32.23 EUR - 46.86 EUR
Product Code | Brand | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Price | Quantity & Availability | ||||||
12302409
|
3M™
7000088756 |
Filter Type: FFABE1P3 R D |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12372399
|
3M™
7000034735 |
Filter Type: FFA1P2 R D |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12392399
|
3M™
7000088755 |
Filter Type: FFA2P3 R D |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12312409
|
3M™
7000088757 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||||
11733343
|
3M™
7000088757 |
Filter Type: FFABEK1P3 R D |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Simple, lightweight, well-balanced respirator offers reliable respiratory protection against a range of gases, vapors, and particulates.- No parts assembly or maintenance required
- Unique bonded carbon technology allows the respirator to sit close to the face
- Low profile design allows the respirator to fit under many other types of protective equipment, providing an undisturbed field of vision and a more comfortable worklife
- Exhalation valve centrally positioned for effective removal of heat and moisture
- Can be used until damaged, particulate filters clog, gas filters become saturated or after one month
- Fine dusts
- Oil and water based mists
- Organic gases or vapours with good warning properties and boiling point above 65°C
- EN 405:2002
- Maximum usage limitations:
- Gases/vapours: APF - 10 x WEL
- Particulates: APF - 20 x WEL
- CE marked