missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark™ Kimtech™ Green Nitrile Ambidextrous Gloves

Kimtech™ Green Nitrile Ambidextrous Gloves offer superb protection with an environmentally-friendly glove thickness of just 0.06 mm. The comfortable design is powder- and latex-free and guards against a number of common contaminants.

Brand:  Kimberly-Clark™ 99853

Additional Details : Weight : 1.33200kg

 View more versions of this product

Product Code. 11749295

  • 46.30 EUR / Pack of 250


Estimated Shipment
25-01-2021
Add to basket

Description

Description

  • Assured Compliance:
    • PPE Cat III according to Regulation (EU) 2016/425
    • EN ISO 374-1:2016 Type C (K) Chemical Splash protection
    • EN 374-4:2003 Resistance to degradation by chemicals
    • EN ISO 374-5:2016 Micro Organism and Virus Protection
  • Nitrile construction results in gloves that are stronger and leaner than latex gloves, and feature better protection against a wider range of contaminants, including viruses, micro-organisms and chemical splash.
  • Gloves are anti-static to protect the wearer and equipment, ambidextrous and the thin construction enables reduced storage space, waste and environmental impact.
  • A textured finish enhances grip and tactile sensitivity for safer and more efficient processes.
  • Beaded cuffs add strength to the gloves, reducing the risk of tearing and increasing their durability, while also reducing roll down for easier donning and doffing.
  • Contain no natural rubber latex, silicone or powder, reducing the risks of skin irritation for the wearer.
  • Food contact approved
Specifications

Specifications

Beaded Cuff
PPE Category III
Nitrile
Large
253 mm
Green
Manufactured in accordance with Quality System ISO 9001, Virus protection according to ISO 16604 procedure B:2004
0.1mm
Beaded/Rolled
Protecting against chemical splash and micro-organism hazards
250/Pk.
Powder-free
250 mm
Videos
SDS
Documents