missing translation for 'onlineSavingsMsg'
Learn More

Diphosgene, 99%, Thermo Scientific Chemicals

Catalog Number p-123100
Click to view available options
Quantity:
10 mL
100 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
503-38-8
C2Cl4O2
197.82
MFCD00015553
HCUYBXPSSCRKRF-UHFFFAOYSA-N
diphosgene, trichloromethyl chloroformate, diphosgen, difosgen czech, perchloromethyl formate, unii-po4q4r80lv, hsdb 371, carbonochloridic acid, trichloromethyl ester, methanol, trichloro-, chloroformate, chloroformic acid trichloromethyl ester
10426
trichloromethyl carbonochloridate
ClC(=O)OC(Cl)(Cl)Cl
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
503-38-8
C2Cl4O2
197.82
MFCD00015553
HCUYBXPSSCRKRF-UHFFFAOYSA-N
diphosgene, trichloromethyl chloroformate, diphosgen, difosgen czech, perchloromethyl formate, unii-po4q4r80lv, hsdb 371, carbonochloridic acid, trichloromethyl ester, methanol, trichloro-, chloroformate, chloroformic acid trichloromethyl ester
10426
trichloromethyl carbonochloridate
ClC(=O)OC(Cl)(Cl)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY
Melting Point -57.0°C
Color Colorless
Density 1.6400g/mL
Boiling Point 128.0°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula ClCOOCCl3
Refractive Index 1.4560 to 1.4590
Quantity 10 mL
Merck Index 15, 3374
Specific Gravity 1.64
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in alcohol, benzene and ether
Formula Weight 197.83
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Diphosgene, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Diphosgene
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Acute toxicity Category 1
Hazard Statement
  • H300-Fatal if swallowed.
  • H330-Fatal if inhaled.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'