missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diphenylphosphine, 95%, Thermo Scientific Chemicals
55.00 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Density | 1.0700 g/mL |
Boiling Point | 280.0°C |
Flash Point | 110°C |
Assay Percent Range | 95% |
Packaging | Glass bottle |
Molecular Formula | C12H11P |
Linear Formula | C6H5PHC6H5 |
Refractive Index | 1.6240 to 1.6260 |
Quantity | 5 g |
Merck Index | 15, 3364 |
Show More |
Safety and Handling
Product Identifier
- Diphenylphosphine
- Danger
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- Pyrophoric liquid Category 1
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- H302-Harmful if swallowed.
- H319-Causes serious eye irritation.
- H250-Catches fire spontaneously if exposed to air.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'