missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(+)-DIOP, 98%, Thermo Scientific™
Click to view available options
Quantity:
0.25 g
1 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
It is employed as a ligand for the in situ preparation of chiral hydrogenation catalysts. It also finds it application for the in situ preparation of chiral hydrogenation catalysts, ligand for Rh(I)-catalyzed asymmetric hydrogenation of enamides, 1,4-disilylation of α,β-unsaturated ketones, Heck reaction of alkenyl iodides, and hydroformylation of olefins.
Solubility
Sparingly soluble in water at 25°C 2.1E-5 g/L.
Notes
Air sensitive. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
It is employed as a ligand for the in situ preparation of chiral hydrogenation catalysts. It also finds it application for the in situ preparation of chiral hydrogenation catalysts, ligand for Rh(I)-catalyzed asymmetric hydrogenation of enamides, 1,4-disilylation of α,β-unsaturated ketones, Heck reaction of alkenyl iodides, and hydroformylation of olefins.
Solubility
Sparingly soluble in water at 25°C 2.1E-5 g/L.
Notes
Air sensitive. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Melting Point | 88°C to 90°C |
Quantity | 0.25 g |
Beilstein | 1630280 |
Sensitivity | Air sensitive |
Solubility Information | Sparingly soluble in water at 25°C 2.1E-5g/L. |
Optical Rotation | +12.5° (c=4.6 in Benzene) |
Formula Weight | 498.55 |
Percent Purity | 98% |
Chemical Name or Material | (+)-DIOP |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'