Learn More
Thermo Scientific™ Dionex™ MAbPac™SCX-10 Ion Exchange Columns
For the high-resolution separation and analysis of monoclonal antibody variants
3799.59 EUR - 3990.00 EUR
Product Code | Brand | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Price | Quantity & Availability | ||||||
15530244
|
Thermo Scientific™
088783 |
Length: 250mm; ID: 4mm; 3 Lot of Resin |
3799.59 EUR
Each |
In Stock
Log in to see stock. |
|||||
15520244
|
Thermo Scientific™
088782 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||||
Description
- Unequalled high-resolution separations
- Exceptionally high efficiency
- Highest resolution of monoclonal antibody variants
- Unmatched column-to-column and lot-to-lot reproducibility
- Hydrophobic interactions eliminated
- Excellent for stability testing
Specifications
Bio-LC |