Learn More
Dimethyl 2-hydroxyethylphosphonate, 95%, Thermo Scientific Chemicals
Catalog Number p-118216
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
54731-72-5
C4H11O4P
154.10
MFCD00043376
TZPPDWDHNIMTDQ-UHFFFAOYSA-N
dimethyl 2-hydroxyethylphosphonate, dimethyl 2-hydroxyethyl phosphonate, 2-hydroxyethylphosphonic acid dimethyl ester, 2-hydroxyethyl phosphonic acid dimethyl ester, acmc-1ao1t, 2-diethylphosphono ethan-1-ol, tzppdwdhnimtdq-uhfffaoysa, hydroxyethyldimethylphosphonic acid, 2-dimethoxyphosphoryl ethan-1-ol
2733833
2-dimethoxyphosphorylethanol
COP(=O)(CCO)OC
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
54731-72-5
C4H11O4P
154.10
MFCD00043376
TZPPDWDHNIMTDQ-UHFFFAOYSA-N
dimethyl 2-hydroxyethylphosphonate, dimethyl 2-hydroxyethyl phosphonate, 2-hydroxyethylphosphonic acid dimethyl ester, 2-hydroxyethyl phosphonic acid dimethyl ester, acmc-1ao1t, 2-diethylphosphono ethan-1-ol, tzppdwdhnimtdq-uhfffaoysa, hydroxyethyldimethylphosphonic acid, 2-dimethoxyphosphoryl ethan-1-ol
2733833
2-dimethoxyphosphorylethanol
COP(=O)(CCO)OC
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 1.2470g/mL
Boiling Point 98.0°C to 104.0°C (0.1 mmHg)
Flash Point 192°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula (CH3O)2P(O)(CH2)2OH
Refractive Index 1.4430 to 1.4460
Quantity 5 mL
Beilstein 04, IV, 3584
Specific Gravity 1.247
Solubility Information Solubility in water: soluble. Other solubilities: soluble in most common organic solvents
Formula Weight 154.1
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Dimethyl 2-hydroxyethylphosphonate, 95%
Show More Show Less
compliance-icons
Product Identifier
  • Dimethyl 2-hydroxyethylphosphonate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.