Learn More
Trajan Scientific and Medical™ Diluter/Dispenser Syringes for Hamilton Microlab 500™
Click to view available options
Volume (Metric):
50 μL
100 μL
250 μL
Unit Size:
Each
Description
Gives better performance and superior quality. Trajan Scientific and Medical Diluter/Dispenser Syringes for Hamilton Microlab 500™ has the knowledge and expertise that goes into manufacturing an SGE syringe.
- High quality solutions for liquid handling
- Accurate and reproducible injections
- Diluter/Dispenser Syringe for Hamilton Microlab 500
Specifications
Specifications
| Description | 60mm stroke, 6-32 UNC thread in plunger button |
| Includes | Plunger stop |
| Length (Metric) | 60 mm |
| Product Type | Hamilton MicroLab 500™ Instrument Syringes |
| Volume (Metric) | 250 μL |
| Connection Type | Luer Lock |