missing translation for 'onlineSavingsMsg'
Learn More

Diethyl pyrazole-3,5-dicarboxylate, 99%, Thermo Scientific Chemicals

Catalog Number 10166683
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
37687-24-4
C9H12N2O4
212.21
MFCD00152167
MBWXLICVQZUJOW-UHFFFAOYSA-N
diethyl 3,5-pyrazoledicarboxylate, diethyl pyrazole-3,5-dicarboxylate, 3,5-diethoxycarbonylpyrazole, 1h-pyrazole-3,5-dicarboxylic acid diethyl ester, 3,5-pyrazoledicarboxylic acid diethyl ester, 3,5-diethyl 1h-pyrazole-3,5-dicarboxylate, 1h-pyrazole-3,5-dicarboxylic acid, diethyl ester, zlchem 861, pubchem22955, acmc-1aj76
142184
diethyl 1H-pyrazole-3,5-dicarboxylate
CCOC(=O)C1=CC(=NN1)C(=O)OCC
This item is not returnable. View return policy
37687-24-4
C9H12N2O4
212.21
MFCD00152167
MBWXLICVQZUJOW-UHFFFAOYSA-N
diethyl 3,5-pyrazoledicarboxylate, diethyl pyrazole-3,5-dicarboxylate, 3,5-diethoxycarbonylpyrazole, 1h-pyrazole-3,5-dicarboxylic acid diethyl ester, 3,5-pyrazoledicarboxylic acid diethyl ester, 3,5-diethyl 1h-pyrazole-3,5-dicarboxylate, 1h-pyrazole-3,5-dicarboxylic acid, diethyl ester, zlchem 861, pubchem22955, acmc-1aj76
142184
diethyl 1H-pyrazole-3,5-dicarboxylate
CCOC(=O)C1=CC(=NN1)C(=O)OCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 51.0°C to 55.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 212.21
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Diethyl pyrazole-3,5-dicarboxylate, 99%
compliance-icons
Product Identifier
  • Diethyl pyrazole-3,5-dicarboxylate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'