missing translation for 'onlineSavingsMsg'
Learn More

Dibromobis(triphenylphosphine)nickel(II), 99%, Thermo Scientific Chemicals

Catalog Number p-7051899
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
14126-37-5
C36H30Br2NiP2
743.09
MFCD00015865
QEKXARSPUFVXIX-UHFFFAOYSA-L
nickel ii bromide bis triphenylphosphine, nibr2 pph3 2, dibromobis triphenylphosphine nickel ii, bis triphenylphosphine nickel ii bromide, dibromobis triphenylphosphine nickel, dibromonickel; bis triphenylphosphine, bis triphenylphosphine dibromonickel ii
11050914
dibromonickel;triphenylphosphane
Br[Ni]Br.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
This item is not returnable. View return policy
14126-37-5
C36H30Br2NiP2
743.09
MFCD00015865
QEKXARSPUFVXIX-UHFFFAOYSA-L
nickel ii bromide bis triphenylphosphine, nibr2 pph3 2, dibromobis triphenylphosphine nickel ii, bis triphenylphosphine nickel ii bromide, dibromobis triphenylphosphine nickel, dibromonickel; bis triphenylphosphine, bis triphenylphosphine dibromonickel ii
11050914
dibromonickel;triphenylphosphane
Br[Ni]Br.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

As a polymerization catalyst

As a polymerization catalyst

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 219°C to 223°C
Odor Odorless
Quantity 1 g
Formula Weight 743.12
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Dibromobis(triphenylphosphine)nickel(II)

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'