Learn More
Diaion™ WK11(H), ion exchange resin, weakly acidic porous type, 2.9 meq/ml on polymethacrylate, Thermo Scientific™
Pharmaceuticals, dealkalination, metal recovery, antibiotics, sucrose purifications
Brand: Thermo Scientific Alfa Aesar 046738.A1
Additional Details : Weight : 1.00000kg
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Beads | |
Weakly acidic porous type, 2.9meq/mL on polymethacrylate | |
White |
Diaion™ WK11 (H), ion exchange resin | |
300 to 1180μm | |
1 kg |