Learn More
Diaion™ PA316(Cl), ion exchange resin, 8% cross-linked, strongly basic porous type I, 1.3 meq/ml on PS-DVB, Thermo Scientific Chemicals
Brand: Thermo Scientific Alfa Aesar 046641.A1
71.33 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Additional Details : Weight : 1.00000kg
Description
Decolorization in sugar solutions, metal recovery, pharmaceuticals, purification of amino acids
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Decolorization in sugar solutions, metal recovery, pharmaceuticals, purification of amino acids
Specifications
300-1180μm Beads | |
8% cross-linked, strongly basic porous type I, 1.3meq/mL on PS-DVB | |
1 kg |
Diaion™ PA316 (Cl), ion exchange resin | |
300 to 1180μm |
RUO – Research Use Only