missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dexamethasone, 99.5%, MP Biomedicals™
Click to view available options
Quantity:
25 mg
100 mg
1 g
Specifications
Color | White |
Assay Percent Range | 0.995 |
Quantity | 25 mg |
Formula Weight | 392.47 |
Physical Form | Powder |
Chemical Name or Material | Dexamethasone, For HPLC analysis |
Safety and Handling
Product Identifier
- Dexamethasone
- Danger
- Reproductive toxicity Category 1B
- H360-May damage fertility or the unborn child.
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : Dexamethasone
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'