Learn More
DAZAP1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA552074
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: EPRDSKSQAP GQPGASQWGS RVVPNAANGW AGQPPPTWQQ GYGPQGMWVP AGQAIGGYGP PPAGRGAPPP PPPFTSYIVS TPPGGFPPPQ GFPQGYGAPP QFSFGYGPPP PPPDQFAPPG VPPPPA Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.
DAZAP1 Polyclonal antibody specifically detects DAZAP1 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
DAZAP1 | |
Polyclonal | |
Unconjugated | |
DAZAP1 | |
2410042M16Rik; DAZ associated protein 1; DAZAP1; DAZ-associated protein 1; Deleted in azoospermia-associated protein 1; LOC561459 protein; LOW QUALITY PROTEIN: DAZ-associated protein 1; MGC19907; mPrrp; testicular tissue protein Li 50 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
26528, 362836, 70248 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96EP5, Q9JII5 | |
DAZAP1 | |
Recombinant protein corresponding to Human DAZAP1 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Research Use Only