missing translation for 'onlineSavingsMsg'
Learn More

D-Nipecotic acid, 96+%, Thermo Scientific Chemicals

Catalog Number p-7038465
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
25137-00-2
C6H11NO2
129.159
MFCD01630787
XJLSEXAGTJCILF-RXMQYKEDSA-N
r-piperidine-3-carboxylic acid, r---nipecotic acid, r-nipecotic acid, 3r-piperidine-3-carboxylic acid, r---3-piperidinecarboxylic acid, --nipecotic acid, d--nipecotic acid, 3r---piperidine-3-carboxylic acid, r-3-piperidinecarboxylic acid, 3r-nipecotic acid
1796510
CHEBI:221278
(3R)-piperidine-3-carboxylic acid
C1CC(CNC1)C(=O)O
This item is not returnable. View return policy
25137-00-2
C6H11NO2
129.159
MFCD01630787
XJLSEXAGTJCILF-RXMQYKEDSA-N
r-piperidine-3-carboxylic acid, r---nipecotic acid, r-nipecotic acid, 3r-piperidine-3-carboxylic acid, r---3-piperidinecarboxylic acid, --nipecotic acid, d--nipecotic acid, 3r---piperidine-3-carboxylic acid, r-3-piperidinecarboxylic acid, 3r-nipecotic acid
1796510
CHEBI:221278
(3R)-piperidine-3-carboxylic acid
C1CC(CNC1)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 250°C to 255°C
Formula Weight 129.16
Percent Purity ≥96%
Quantity 1 g
Optical Rotation −5° (c=1 in Water)
Chemical Name or Material D-Nipecotic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'