missing translation for 'onlineSavingsMsg'
Learn More

D-Biotin, ≥97.5%, Cell Culture Reagent Grade, MP Biomedicals™

Catalog Number p-4405717
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
58-85-5
C10H16N2O3S
244.31
MFCD00005541
YBJHBAHKTGYVGT-UHFFFAOYNA-N
biotin, d-biotin, vitamin h, coenzyme r, vitamin b7, bios ii, factor s, bioepiderm, d +-biotin, biodermatin
171548
CHEBI:15956
OC(=O)CCCCC1SCC2NC(=O)NC12
This item is not returnable. View return policy
58-85-5
C10H16N2O3S
244.31
MFCD00005541
YBJHBAHKTGYVGT-UHFFFAOYNA-N
biotin, d-biotin, vitamin h, coenzyme r, vitamin b7, bios ii, factor s, bioepiderm, d +-biotin, biodermatin
171548
CHEBI:15956
OC(=O)CCCCC1SCC2NC(=O)NC12
Color White
Density 1.267 g/cm3(Lit.)
Assay Percent Range ≥97.5%
Quantity 1 g
IUPAC Name 5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]pentanoic acid
Formula Weight 244.3
Grade Cell Culture Reagent
Physical Form Crystalline Powder
Chemical Name or Material D-Biotin
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'