Learn More
Thermo Scientific™ D.A.R.T.'S™ (Disposable Automation Research Tips)
D.A.R.T.«S (Disposable Automation Research Tips) have been designed for the ultimate performance in automated pipetting
Brand: Thermo Scientific™ 5518-11
417.00 EUR valid until 2024-12-31
Use promo code "21898" to get your promotional price.
Additional Details : Weight : 0.21000kg
Includes: 20 magazines per case
Description
Unlike conventional automation tips, D.A.R.T. tips are provided in disposable rigid magazines that seal onto the pipetting heads by sealing against a silicone pad. This forms a definitive seal which minimizes tip-to-tip height variations and elimiates static interactions, enabling consistent dispensing into the bottom of microplate wells.
- Compatible with the Matrix Hydra DT and Matrix PlateMate 2x3 and PlateMate Plus Systems
- Stepped-body design minimizes volume displacement and ensures tip straightness for better well access
- Highly polished molds and cores ensure low liquid retention
- Small internal tip orifice provides better small volume droplet formation, resulting in optimal accuracy and precision
- Thorough inspection for proper formation as well as straightness ensures tips are free of defects
- Unique face seal design ensures consistent tip-to-tip height for precise low volume dispensing
- Entire assembly can be quickly removed and replaced, eliminating extra handling of the tips which can cause damage or contamination
Specifications
300 μL | |
Automation | |
Thermo Scientific Versette, Thermo Scientific Matrix PlateMate 2x3, Thermo Scientific Matrix Hydra DT |
Sterile | |
Single-use | |
D.A.R.T.S |