missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 6 Mouse, Clone: KRT6/1702, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 3853-MSM2-P1
This item is not returnable.
View return policy
Specifications
Cytokeratin 6 | |
Cytoplasmic | |
KRT6/1702 | |
Unconjugated | |
NP_775109 | |
Mouse | |
56kDa | |
Protein A/G Purified | |
RUO | |
286887 | |
IgG2a κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
KRT6A ;KRT6B ;KRT6C | |
Recombinant human full-length keratin 6 protein | |
Tonsil or basal cell carcinoma | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |