missing translation for 'onlineSavingsMsg'
Learn More

Cytidine-5'-diphosphate trisodium salt, Thermo Scientific™

Catalog Number p-7042917
Click to view available options
Quantity:
100 mg
500 mg
1 g
This item is not returnable. View return policy
34393-59-4
C9H12N3Na3O11P2
469.12
MFCD00065201
JENUKVZGXGULDX-LLWADOMFSA-K
cytidine 5'-diphosphate trisodium salt
131674171
[(2R,3S,4R,5R)-5-(4-amino-2-oxopyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphono hydrogen phosphate;molecular hydrogen;sodium
[Na+].[Na+].[Na+].NC1=NC(=O)N(C=C1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])([O-])=O)[C@@H](O)[C@H]1O
This item is not returnable. View return policy
34393-59-4
C9H12N3Na3O11P2
469.12
MFCD00065201
JENUKVZGXGULDX-LLWADOMFSA-K
cytidine 5'-diphosphate trisodium salt
131674171
[(2R,3S,4R,5R)-5-(4-amino-2-oxopyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphono hydrogen phosphate;molecular hydrogen;sodium
[Na+].[Na+].[Na+].NC1=NC(=O)N(C=C1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])([O-])=O)[C@@H](O)[C@H]1O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Cytidine-5′-diphosphate trisodium salt has been used to produce a vaccinia virus that induces ribonucleotide reductase in primate cells.

Solubility
Freely soluble in water, insoluble in ethanol, chloroform and ether.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
TRUSTED_SUSTAINABILITY
Color White
Quantity 100 mg
Solubility Information Freely soluble in water,insoluble in ethanol,chloroform and ether.
Formula Weight 469.12
Physical Form Crystalline Powder
Chemical Name or Material Cytidine-5'-diphosphate trisodium salt
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'