Learn More
Lenz Laborglasinstrumente™ Cylindrical Dropping Funnels
For the safe, easy and accurate transfer of fluids in the laboratory. Lenz Laborglasinstrumente™ Cylindrical Dropping Funnels are manufactured from durable DURAN™ borosilicate glass.
Brand: Lenz Laborglasinstrumente™ B0496701
Additional Details : Weight : 0.14100kg
Description
- With inscription label
- With NS socket and cone
- With scale (approx. volume/mL)
- With pressure equalizing tube
- With screwthread retaining device
- Corrosion-free and resistant to thermal shock
Specifications
50 mL | |
Duran Glass, Borosilicate Glass | |
Pressure Equalization Tube at the Side |