Learn More
Macherey-Nagel™ CW Deactivated Capillary Column
Brand: Macherey-Nagel™ 723349.10
Additional Details : Weight : 0.01000kg
Specifications
0.32 mm | |
0.5 mm | |
250°C | |
1 Each |
Deactivated Capillary Column | |
Preparation of Capillary Columns Analysis | |
10 m |