Learn More
Invitrogen™ Custom Silencer™ siRNA
RNA interference (RNAi) is the best way to effectively knock down gene expression to study protein function in a wide range of cell types.
Brand: Invitrogen™ AM16233
9453.70 EUR valid until 2024-06-28
Use promo code "21649" to get your promotional price.
Additional Details : Weight : 0.00450kg
Description
Use our GeneAssist™ Custom siRNA Builder to design and order a custom siRNA.
Specifications
Store at or below -20°C. Do not store in a frost-free freezer. Dried oligonucleotides are shipped at ambient temperature. | |
siRNA | |
Ambion™, Silencer™ Select | |
10 μmol |
Custom siRNA, 10 umol | |
RNA | |
siRNA |
For Research Use Only. Not for use in diagnostic procedures.