Learn More
Thermo Scientific™ Culti-Loops™ Citrobacter diversus KM 11012
Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Brand:  Thermo Scientific™ R4601508
Additional Details : Weight : 0.40000kg Transport : UN number : 3373 Chem class : 6.2
Product Code. 12958898
  • 189.00 EUR / Pack of 5
Estimated Shipment: 08-05-2024
Description
Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.
Ordering Information:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
Specifications
Specifications
Citrobacter species
Citrobacter diversus KM 11012
Gram-negative Rods
5 Loops/Pack
KM 11012TM*
Culti-Loops Quality Control Organism
Qualitative QC
Special Offers
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.