missing translation for 'onlineSavingsMsg'
Learn More

CSNK1G3 (CK1 γ 3) Recombinant Human Protein

CSNK1G3 (CK1 γ 3) Recombinant Human Protein

Brand:  Invitrogen PR7474A

Additional Details : Weight : 0.00001kg

 View more versions of this product

Product Code. 15485030

  • 1826.00 EUR / 100µg
Estimated Shipment: 18-04-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The casein kinase I (CKI) gene family encodes serine⁄threonine kinases that preferentially phosphorylate acidic substrates. CKI proteins are monomeric and range from 25 to 55 kD. They are ubiquitous, being found in the nuclei, cytoplasm, and membrane fractions of eukaryotic cells
Specifications

Specifications

NP_004375, R174G
Kinase Assay
78.9kDa
100μg
For research use only. Not for use in diagnostic procedures
CSNK1G3 (CK1 γ 3)
CSNK1G3 was pre-diluted in enzyme dilution buffer and assayed in 25mM Tris (pH 7.5), 10mM MgCl2, 0.5mM EGTA, 0.5mM Na3VO4, 5mM β-glycerophosphate, 2.5mM DTT, 0.01% Triton™ X-100, 200μM ATP, 330μg/mL casein kinase 1 peptide substrate (RRKDLHDDEEDEAMSITA) and trace [32P]-γ-ATP for 10 minutes at 30°C
CSNK1G3 (CK1 γ 3) Recombinant Human Protein
Human
Baculovirus
80% by SDS-PAGE
0.36mg/mL
Frozen
CSNK1G3 (CK1 γ 3) Recombinant Human Protein
–80°C
1 Each
CSNK1G3 (CK1 γ 3) Recombinant Human Protein
CK1
160nmoles of phosphate transferred to Casein Kinase 1 peptide substrate (RRKDLHDDEEDEAMSITA) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 3.33μg/mL
GST Tag
Frozen
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.